Learn More
Medchemexpress LLC 2-(Trifluoromethyl)cinnamic acid | 2062-25-1 | 99.8% | C10H7F3O2 | 5 G
SDP
Catalog No. 5000434315
Encompass_Preferred
2-(Trifluoromethyl)cinnamic acid is a cinnamic acid derivative that inhibits the proton pump (H+/K+-ATPase), reducing gastric acid secretion. It also improves delayed gastric emptying and can be used in research on gastric diseases such as acute gastritis and gastric ulcers.
  • Cinnamic acid derivative
  • Inhibits proton pump (H+/K+-ATPase)
  • Reduces gastric acid secretion
  • Improves delayed gastric emptying
  • Suitable for research on gastric diseases
  • White to off-white solid appearance
Catalog No. 50-004-34315 Supplier Medchemexpress LLC Supplier No. HY75424A5G
2-(Trifluoromethyl)cinnamic acid is a cinnamic acid derivative that inhibits the proton pump (H+/K+-ATPase), reducing gastric acid secretion. It also improves delayed gastric emptying and can be used in research on gastric diseases such as acute gastritis and gastric ulcers.
  • Cinnamic acid derivative
  • Inhibits proton pump (H+/K+-ATPase)
  • Reduces gastric acid secretion
  • Improves delayed gastric emptying
  • Suitable for research on gastric diseases
  • White to off-white solid appearance