Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Trifluoromethyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. T18005G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1800-5G 5 g
T1800-25G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T1800-5G Supplier TCI America Supplier No. T18005G
Add to Cart
Add to Cart

Specifications

CAS 1423-27-4
Color White-Yellow
Molecular Formula C7H6BF3O2
Quantity 5 g
MDL Number MFCD00236059
Synonym 2-trifluoromethyl phenylboronic acid, 2-trifluoromethylphenylboronic acid, 2-trifluoromethyl phenyl boronic acid, 2-trifluoromethyl benzeneboronic acid, 2-trifluoromethyl phenyl boranediol
InChI Key JNSBEPKGFVENFS-UHFFFAOYSA-N
SMILES B(C1=CC=CC=C1C(F)(F)F)(O)O
IUPAC Name [2-(trifluoromethyl)phenyl]boronic acid
Molecular Weight (g/mol) 189.928
PubChem CID 2734387
Formula Weight 189.93
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.