Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+/-)-2-(Trifluoromethyl)piperazine, 97%, Thermo Scientific Chemicals

Catalog No. AAH32524MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH32524MD 250 mg
AAH3252403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH32524MD Supplier Thermo Scientific Chemicals Supplier No. H32524MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 131922-05-9 | Molecular Formula: C5H9F3N2 | Molecular Weight: 154.136 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131922-05-9
Molecular Formula C5H9F3N2
Molecular Weight (g/mol) 154.136
MDL Number MFCD07373382
InChI Key YNEJOOCZWXGXDU-UHFFFAOYSA-N
Synonym 2-trifluoromethyl piperazine, 2-trifluoromethyl-piperazine, +/--2-trifluoromethyl piperazine, piperazine, 2-trifluoromethyl, -2-trifluoromethyl piperazine, piperazine, 2-trifluoromethyl-9ci, acmc-20aolo, 2trifluoromethylpiperazine, m-trifluoromethylpiperazine
PubChem CID 18519725
IUPAC Name 2-(trifluoromethyl)piperazine
SMILES C1CNC(CN1)C(F)(F)F

Specifications

Melting Point 94°C to 97°C
Quantity 250 mg
UN Number UN3259
Sensitivity Air Sensitive
Formula Weight 154.13
Percent Purity 97%
Chemical Name or Material (+/-)-2-(Trifluoromethyl)piperazine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.