Learn More
2-(Trimethylsiloxy)ethyl methacrylate, 94%, stab. with 100ppm 4-methoxyphenol, Thermo Scientific Chemicals
Description
2-(Trimethylsiloxy)ethyl methacrylate is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-(Trimethylsiloxy)ethyl methacrylate is used as pharmaceutical intermediates.
Solubility
Difficult to mix.
Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
| Density | 0.995 |
| Boiling Point | 65°C (0.9 mmHg) |
| Flash Point | 42°C (107°F) |
| Refractive Index | 1.43 |
| Quantity | 50 g |
| Sensitivity | Moisture sensitive |
| Solubility Information | Difficult to mix. |
| Formula Weight | 202.33 |
| Percent Purity | 94% |
| Chemical Name or Material | 2-(Trimethylsiloxy)ethyl methacrylate, stabilized with 100ppm 4-methoxyphenol |
Safety and Handling
| Hazard Category | H227-H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335-H227 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. Combustible liquid. |
| Precautionary Statement | P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c |
| EINECSNumber | 241-432-1 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.