Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-(Trimethylsilyl)Ethanesulfonyl Chloride

Catalog No. 6813345G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
68-133-45G 5 g
68-133-41G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 68-133-45G Supplier Sigma Aldrich Supplier No. 6813345G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reagent used for protection of an amino group as a SES-amide. The SES group can be conveniently removed by fluoride ion.

Specifications

CAS 106018-85-3
Density 1.059 g/mL at 25°C
Boiling Point 146.8°C (760 mmHg)
Molecular Formula C5H13ClO2SSi
Refractive Index n20/D 1.4444
Linear Formula (CH3)3SiCH2CH2SO2Cl
MDL Number MFCD09265153
Quantity 5 g
Synonym 2-Trimethylsilylethylsulfonyl chloride; SES-Cl
Molecular Weight (g/mol) 200.76 g/mol
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.