Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-(Trimethylsilyl)Ethanol

Catalog No. 22689050G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-689-050G 50 g
22-689-01G 1 g
22-689-010G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Warning
Catalog No. 22-689-050G Supplier Sigma Aldrich Supplier No. 22689050G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Used to synthesize Teoc-protected amines via alcoholysis of the corresponding isocyanates.

Specifications

CAS 2916-68-9
Density 0.825 g/mL at 25°C
Boiling Point 71°C to 73°C35 mmHg, lit.)
Molecular Formula C5H14OSi
Refractive Index n20/D 1.423
Linear Formula (CH3)3SiCH2CH2OH
MDL Number MFCD00002825
Quantity 50 g
Synonym (2-Hydroxyethyl)trimethylsilane
Molecular Weight (g/mol) 118.25 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.