Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-(Trimethylsilyl)Ethoxymethyl Chloride

Catalog No. 9274925ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
92-749-25ML 25 mL
92-749-5ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 92-749-25ML Supplier Sigma Aldrich Supplier No. 9274925ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-(Trimethylsilyl)ethoxymethyl chloride (SEM-Cl) is a widely used reagent for the preparation protection groups for amines, alcohols, phenols, and carboxy groups. The nitrogen of amides and sulfonamides groups are also protected by using SEM-Cl reagent.SEM-Cl can be used as a reagent for the preparation of 2,4,5-trisubstituted tetrahydropyrans form homoallylic alcohols by a Prins-type cyclization reaction. It can also be used in the synthesis of amino acid SEM esters from N-protected amino acids in the presence of lithium carbonate.Protecting-group reagent for alcohols and other functional groups selectively removable with Bu4NF ; Protecting group for pyrroles and imidazoles, facilitating selective metalation ; Reagent for the introduction of a protected C1 building block

Specifications

CAS 76513-69-4
Density 0.942 g/mL at 25°C
Boiling Point 170°C to 172°C, 57°C to 59°C (8 mmHg, lit.)
Molecular Formula C6H15ClOSi
Refractive Index n20/D 1.435; n20/D 1.436
Linear Formula (CH3)3SiCH2CH2OCH2Cl
MDL Number MFCD00009919
Quantity 25 mL
Synonym (2-Chloromethoxyethyl)trimethylsilane; Chloromethyl 2-trimethylsilylethyl ether; SEM-Cl; SEM-chloride
Molecular Weight (g/mol) 166.72 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.