Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-(Trimethylsilyl)Ethoxymethyl Chloride

Catalog No. 2389025G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-890-25G 5 g
23-890-225G 25 g
23-890-2100G 100 g
23-890-21G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Danger
Catalog No. 23-890-25G Supplier Sigma Aldrich Supplier No. 2389025G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Learn More at the Professor and Product Portal of Professor Bruce Lipshutz.Used to prepare a SEM-ether which was, in turn, removed with BBr3 and cyclized to a benzo-oxapane.

Specifications

CAS 76513-69-4
Density 0.942 g/mL at 25°C
Boiling Point 170°C to 172°C, 57°C to 59°C (8 mmHg, lit.)
Molecular Formula C6H15ClOSi
Refractive Index n20/D 1.435
Linear Formula (CH3)3SiCH2CH2OCH2Cl
MDL Number MFCD00009919
Quantity 5 g
Synonym (2-Chloromethoxyethyl)trimethylsilane; Chloromethyl 2-trimethylsilylethyl ether; SEM-Cl; SEM-chloride
Molecular Weight (g/mol) 166.72 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.