Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Trimethylsilylethynyl)thiophene 98.0+%, TCI America™
SDP

Catalog No. T31891G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T31891G 1 g
T31895G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T31891G Supplier TCI America Supplier No. T31891G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 40231-03-6
Molecular Formula C9H12SSi
Molecular Weight (g/mol) 180.34
MDL Number MFCD03427125
InChI Key OQUBLKNISPLGJP-UHFFFAOYSA-N
Synonym Trimethyl(thiophen-2-ylethynyl)silane
PubChem CID 596870
IUPAC Name trimethyl[2-(thiophen-2-yl)ethynyl]silane
SMILES C[Si](C)(C)C#CC1=CC=CS1

Specifications

Color Yellow
Boiling Point 90°C
Quantity 1 g
UN Number 1993
Formula Weight 180.34
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(Trimethylsilylethynyl)thiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.