Learn More
Cayman Chemical 2-trns-HexadecenoIc AcId 100mg
SDP
Catalog No. 502951354
Encompass_Preferred
An intermediate formed in the β-oxidation of palmitic acid
Catalog No. 50-295-1354 Supplier Cayman Chemical Supplier No. 11132100 MG
An intermediate formed in the β-oxidation of palmitic acid