Learn More
Cayman Chemical Δ2-trans-Hexadecenoic Acid, Purity: ≥95%, CAS: 929-79-3, 50 mg
SDP
Catalog No. 502951418
Encompass_Preferred
An intermediate formed in the β-oxidation of palmitic acid
Catalog No. 50-295-1418 Supplier Cayman Chemical Supplier No. 1113250 MG
An intermediate formed in the β-oxidation of palmitic acid