Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Undecanol 5g | 1653-30-1 | 172.31 | C11H24O | 5 G
SDP

Catalog No. 5000226143
Encompass_Preferred

2-Undecanol (undecan-2-ol; CAS 1653-30-1) is a medium-chain secondary alcohol used as a volatile organic compound and pheromone component in research. It is supplied as a colorless to light yellow liquid and is commonly provided in small laboratory pack sizes for analytical and synthetic applications.

  • High purity (~98%) suitable for research use
  • Molecular formula C11H24O; molecular weight 172.31 g/mol
  • Available in small laboratory pack sizes, including 5 g
  • Used in analytical, synthetic, and pheromone studies
  • Liquid at room temperature; density approximately 0.827 g/cm3

Catalog No. 50-002-26143 Supplier Medchemexpress LLC Supplier No. HY1156845G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Undecanol (undecan-2-ol; CAS 1653-30-1) is a medium-chain secondary alcohol used as a volatile organic compound and pheromone component in research. It is supplied as a colorless to light yellow liquid and is commonly provided in small laboratory pack sizes for analytical and synthetic applications.

  • High purity (~98%) suitable for research use
  • Molecular formula C11H24O; molecular weight 172.31 g/mol
  • Available in small laboratory pack sizes, including 5 g
  • Used in analytical, synthetic, and pheromone studies
  • Liquid at room temperature; density approximately 0.827 g/cm3

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.