Learn More
Medchemexpress LLC 2-Undecanone | 112-12-9 | MFCD00009583 | 99.5% | 170.29 g/mol | C11H22O | 1 ML
SDP
Catalog No. 5000301059
Encompass_Preferred
2-Undecanone is a volatile aliphatic ketone (CAS 112-12-9) that inhibits the DnaKJE-ClpB bichaperone-dependent refolding of heat-inactivated bacterial luciferases and has reported activity against lung tumorigenesis. It is supplied as a ready-to-use 10 mM solution in DMSO and is also available as neat liquid in multiple sizes.
  • Inhibits DnaKJE-ClpB bichaperone-dependent refolding of heat-inactivated bacterial luciferases.
  • Reported to inhibit lung tumorigenesis.
  • Volatile aliphatic ketone with molecular formula C11H22O.
  • Offered as a ready-to-use 10 mM solution in DMSO and as neat liquid sizes for formulation flexibility.
  • High purity suitable for biochemical and cellular assays.
Catalog No. 50-003-01059 Supplier Medchemexpress LLC Supplier No. HYW01696910MM/1ML
2-Undecanone is a volatile aliphatic ketone (CAS 112-12-9) that inhibits the DnaKJE-ClpB bichaperone-dependent refolding of heat-inactivated bacterial luciferases and has reported activity against lung tumorigenesis. It is supplied as a ready-to-use 10 mM solution in DMSO and is also available as neat liquid in multiple sizes.
  • Inhibits DnaKJE-ClpB bichaperone-dependent refolding of heat-inactivated bacterial luciferases.
  • Reported to inhibit lung tumorigenesis.
  • Volatile aliphatic ketone with molecular formula C11H22O.
  • Offered as a ready-to-use 10 mM solution in DMSO and as neat liquid sizes for formulation flexibility.
  • High purity suitable for biochemical and cellular assays.