Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-undecanone | 112-12-9 | MFCD00009583 | 99.5% | 170.29 g/mol | C11H22O | 50 G
SDP

Catalog No. 5000299991
Encompass_Preferred

2-Undecanone (undecan-2-one; CAS 112-12-9) is a volatile aliphatic ketone commonly used as a research reagent and reference standard. It has the molecular formula C11H22O and molecular weight 170.29 g/mol. Reported biological activities in research literature include inhibition of the DnaKJE-ClpB bichaperone-dependent refolding of heat-inactivated bacterial luciferases and reported effects on lung tumorigenesis.

  • Volatile aliphatic ketone with fruity, waxy odor.
  • Molecular formula C11H22O and molecular weight 170.29 g/mol.
  • High purity (≈99.5%) suitable for analytical and research use.
  • Applicable as a biochemical assay reagent and reference standard.
  • Soluble in common organic solvents; follow the safety data sheet for handling and storage.

Catalog No. 50-002-99991 Supplier Medchemexpress LLC Supplier No. HYW01696950G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Undecanone (undecan-2-one; CAS 112-12-9) is a volatile aliphatic ketone commonly used as a research reagent and reference standard. It has the molecular formula C11H22O and molecular weight 170.29 g/mol. Reported biological activities in research literature include inhibition of the DnaKJE-ClpB bichaperone-dependent refolding of heat-inactivated bacterial luciferases and reported effects on lung tumorigenesis.

  • Volatile aliphatic ketone with fruity, waxy odor.
  • Molecular formula C11H22O and molecular weight 170.29 g/mol.
  • High purity (≈99.5%) suitable for analytical and research use.
  • Applicable as a biochemical assay reagent and reference standard.
  • Soluble in common organic solvents; follow the safety data sheet for handling and storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.