Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-undecanone | 112-12-9 | MFCD00009583 | 99.5% | 170.30 g/mol | C11H22O | 1 G
SDP

Catalog No. 5000300597
Encompass_Preferred

2-Undecanone (CAS 112-12-9) is a volatile organic ketone used in research that inhibits the DnaKJE-ClpB bichaperone-dependent refolding of heat-inactivated bacterial luciferases and has reported activity against lung tumorigenesis. It is supplied as a liquid and can be prepared in various solvents for in vitro and in vivo experiments.

  • High purity ≈99.5%.
  • Chemical formula C11H22O; molecular weight 170.30 g/mol.
  • Available in solid and solution formats, e.g., 1 g and 10 mM in DMSO.
  • Colorless to light yellow liquid; density 0.825 g/cm³.
  • Storage: store dry and cool; stock solutions -80°C (2 years), -20°C (1 year).
  • For research use only; not for human therapeutic use.

Catalog No. 50-003-00597 Supplier Medchemexpress LLC Supplier No. HYW0169691G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Undecanone (CAS 112-12-9) is a volatile organic ketone used in research that inhibits the DnaKJE-ClpB bichaperone-dependent refolding of heat-inactivated bacterial luciferases and has reported activity against lung tumorigenesis. It is supplied as a liquid and can be prepared in various solvents for in vitro and in vivo experiments.

  • High purity ≈99.5%.
  • Chemical formula C11H22O; molecular weight 170.30 g/mol.
  • Available in solid and solution formats, e.g., 1 g and 10 mM in DMSO.
  • Colorless to light yellow liquid; density 0.825 g/cm³.
  • Storage: store dry and cool; stock solutions -80°C (2 years), -20°C (1 year).
  • For research use only; not for human therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.