Learn More
Medchemexpress LLC 2-undecanone | 112-12-9 | MFCD00009583 | 99.5% | 170.30 g·mol⁻¹ | C11H22O | 5 G
SDP
Catalog No. 5000300433
Encompass_Preferred
2-Undecanone (CAS 112-12-9) is a small-molecule ketone used as a research reagent and analytical standard. It is a volatile organic compound reported to inhibit DnaKJE-ClpB-dependent refolding of heat-inactivated bacterial luciferases and has been investigated in lung tumorigenesis studies. Supplied in laboratory-scale quantities for research use.
  • Volatile organic ketone suitable for research applications.
  • Reported inhibitor of DnaKJE-ClpB chaperone-dependent refolding.
  • Provided in multiple laboratory-scale quantities for flexibility.
  • High purity for analytical and experimental use.
  • Molecular formula C11H22O; molecular weight 170.30 g·mol⁻¹.
Catalog No. 50-003-00433 Supplier Medchemexpress LLC Supplier No. HYW0169695G
2-Undecanone (CAS 112-12-9) is a small-molecule ketone used as a research reagent and analytical standard. It is a volatile organic compound reported to inhibit DnaKJE-ClpB-dependent refolding of heat-inactivated bacterial luciferases and has been investigated in lung tumorigenesis studies. Supplied in laboratory-scale quantities for research use.
  • Volatile organic ketone suitable for research applications.
  • Reported inhibitor of DnaKJE-ClpB chaperone-dependent refolding.
  • Provided in multiple laboratory-scale quantities for flexibility.
  • High purity for analytical and experimental use.
  • Molecular formula C11H22O; molecular weight 170.30 g·mol⁻¹.