Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Undecanone, 98%

Catalog No. p-210557
Change view
Click to view available options
Quantity:
5 mL
250 mL
1 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC140690050 5 mL Glass bottle
AC140692500 250 mL Glass bottle
AC140690010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC140690050 Supplier Thermo Scientific Chemicals Supplier No. 140690050
Only null left
Add to Cart

CAS: 112-12-9 | C11H22O | 170.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-12-9
Molecular Formula C11H22O
Molecular Weight (g/mol) 170.30
MDL Number MFCD00009583
InChI Key KYWIYKKSMDLRDC-UHFFFAOYSA-N
Synonym 2-undecanone, methyl nonyl ketone, 2-hendecanone, ketone, methyl nonyl, undecanone, nonyl methyl ketone, rue ketone, 2-oxoundecane, methyl n-nonyl ketone, methylnonylketone
PubChem CID 8163
ChEBI CHEBI:17700
IUPAC Name undecan-2-one
SMILES CCCCCCCCCC(C)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 11.0°C to 13.0°C
Color Colorless to Yellow
Density 0.8240g/mL
Boiling Point 231.0°C to 232.0°C
Flash Point 88°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4280 to 1.4310
Linear Formula CH3(CH2)8COCH3
Quantity 5 mL
Beilstein 01, 713
Specific Gravity 0.824
Merck Index 15, 6177
Solubility Information Solubility in water: insoluble
Formula Weight 170.3
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Undecanone, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Avoid release to the environment.
Collect spillage.
Dispose of contents/container to an approved waste disposal plant.
EINECSNumber 203-937-5
RTECSNumber YQ2820000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.