Learn More
Medchemexpress LLC 2-undecanone (methyl nonyl ketone) | 112-12-9 | MFCD00009583 | 99.5% | 170.30 g/mol | C11H22O | 10 G
SDP
Catalog No. 5000299880
Encompass_Preferred
2-Undecanone (methyl nonyl ketone) is a volatile organic ketone used as a biochemical reagent in research. It has reported activity inhibiting the DnaKJE-ClpB bichaperone-dependent refolding of heat-inactivated bacterial luciferases and has been reported to reduce lung tumorigenesis in experimental studies. It is supplied at high purity for analytical and research use.
  • Volatile organic ketone used as a research reagent.
  • Inhibits DnaKJE-ClpB-dependent refolding of heat-inactivated bacterial luciferases.
  • Reported to inhibit lung tumorigenesis in experimental models.
  • High purity suitable for analytical and research applications.
  • Available in multiple pack sizes, including small laboratory quantities.
Catalog No. 50-002-99880 Supplier Medchemexpress LLC Supplier No. HYW01696910G
2-Undecanone (methyl nonyl ketone) is a volatile organic ketone used as a biochemical reagent in research. It has reported activity inhibiting the DnaKJE-ClpB bichaperone-dependent refolding of heat-inactivated bacterial luciferases and has been reported to reduce lung tumorigenesis in experimental studies. It is supplied at high purity for analytical and research use.
  • Volatile organic ketone used as a research reagent.
  • Inhibits DnaKJE-ClpB-dependent refolding of heat-inactivated bacterial luciferases.
  • Reported to inhibit lung tumorigenesis in experimental models.
  • High purity suitable for analytical and research applications.
  • Available in multiple pack sizes, including small laboratory quantities.