Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Undecen-1-al, MP Biomedicals

Catalog No. ICN218570 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN218570 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN218570 Supplier MP Biomedicals, Inc Supplier No. 0521857080

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2463-77-6
Molecular Formula C11H20O
Molecular Weight (g/mol) 168.28
InChI Key PANBRUWVURLWGY-MDZDMXLPSA-N
Synonym trans-2-undecenal, 2-undecenal, e-undec-2-enal, e-2-undecenal, 2-undecenal, 2e, 2-undecen-1-al, 2e-2-undecenal, trans-2-undecen-1-al, 2-undecenal, e, unii-fnp3s9mg30
PubChem CID 5283356
IUPAC Name (E)-undec-2-enal
SMILES CCCCCCCCC=CC=O

Specifications

Quantity 100 g
Formula Weight 168
Chemical Name or Material 2-Undecen-1-Al
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.