Learn More
Medchemexpress LLC 2-undecyl-4R-thiazolidinecarboxylic acid | 298186-80-8 | ≥98.0% | 5 MG
SDP
Catalog No. 5000234027
Encompass_Preferred
CAY10444 (BML-241) is a small-molecule antagonist of the sphingosine-1-phosphate receptor 3 (S1P3), used as a research reagent to study S1P3-mediated calcium mobilization and related cellular responses in vitro.
  • Antagonist of S1P3, used in cell-based calcium and migration assays.
  • High purity: ≥98.0%.
  • Molecular weight 287.5 g/mol; formula C15H29NO2S.
  • Crystalline solid; store at -20°C.
  • Supplied as a small research vial (typical single-vial quantity: 5 mg).
Catalog No. 50-002-34027 Supplier Medchemexpress LLC Supplier No. HY1194015MG
CAY10444 (BML-241) is a small-molecule antagonist of the sphingosine-1-phosphate receptor 3 (S1P3), used as a research reagent to study S1P3-mediated calcium mobilization and related cellular responses in vitro.
  • Antagonist of S1P3, used in cell-based calcium and migration assays.
  • High purity: ≥98.0%.
  • Molecular weight 287.5 g/mol; formula C15H29NO2S.
  • Crystalline solid; store at -20°C.
  • Supplied as a small research vial (typical single-vial quantity: 5 mg).