Learn More
OHAUS 2 Universal Clip Plates
Catalog No. 01804332 Shop All OHAUS Products
Change view
Click to view available options
Product Type:
Shaker Attachment
Catalog No. Product Type
01-804-332 Shaker Attachment
1 options
Catalog No. 01-804-332 Supplier OHAUS Supplier No. 83041374
Add to Cart
Add to Cart
Pack of 2 clip plates with two elastic bands each for use with odd-shapped sample vessels.
  • This can be attached to the Magnetic Platform for use with the Roto-Shake Genie, Rotator Genie, Enviro-Genie or Incubator-Genie.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Shaker Attachment