Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Vinylphenyl acetate | 63600-35-1 | 98.4% | 162.19 | 1 G
SDP

Catalog No. 5000400881
Encompass_Preferred

2-Vinylphenyl acetate is a chemical compound primarily utilized as a laboratory chemical and in the manufacture of various substances. It appears as a colorless to light yellow liquid with a molecular weight of 162.19.

  • Used as a laboratory chemical
  • Employed in the manufacture of substances
  • Liquid appearance
  • Purity of 98.37%
  • Store at 4°C, protected from light and under nitrogen

Catalog No. 50-004-00881 Supplier Medchemexpress LLC Supplier No. HY239251G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Vinylphenyl acetate is a chemical compound primarily utilized as a laboratory chemical and in the manufacture of various substances. It appears as a colorless to light yellow liquid with a molecular weight of 162.19.

  • Used as a laboratory chemical
  • Employed in the manufacture of substances
  • Liquid appearance
  • Purity of 98.37%
  • Store at 4°C, protected from light and under nitrogen

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.