Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Vinylphenyl acetate | 63600-35-1 | 98.4% | 162.19 | 50 G
SDP

Catalog No. 5000402143
Encompass_Preferred

This product, 2-Vinylphenyl acetate, is a chemical intended for laboratory use. It is classified under GHS as causing skin and serious eye irritation, requiring careful handling with appropriate personal protective equipment. It is recommended for storage at 4°C, protected from light, and under nitrogen atmosphere. This chemical appears as a colorless to light yellow liquid.

  • Causes skin irritation
  • Causes serious eye irritation
  • Store at 4°C, protected from light, and under nitrogen
  • Appearance: colorless to light yellow liquid
  • Molecular formula: C10H10O2
  • Molecular weight: 162.19
  • Purity: 98.4%

Catalog No. 50-004-02143 Supplier Medchemexpress LLC Supplier No. HY2392550G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product, 2-Vinylphenyl acetate, is a chemical intended for laboratory use. It is classified under GHS as causing skin and serious eye irritation, requiring careful handling with appropriate personal protective equipment. It is recommended for storage at 4°C, protected from light, and under nitrogen atmosphere. This chemical appears as a colorless to light yellow liquid.

  • Causes skin irritation
  • Causes serious eye irritation
  • Store at 4°C, protected from light, and under nitrogen
  • Appearance: colorless to light yellow liquid
  • Molecular formula: C10H10O2
  • Molecular weight: 162.19
  • Purity: 98.4%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.