Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Vinylpyridine

Catalog No. 1322925ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-229-25ML 5 mL
13-229-2100ML 100 mL
13-229-2500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 13-229-25ML Supplier Sigma Aldrich Supplier No. 1322925ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Vinylpyridine (2VP) is a water soluble pyridine derivative that can be used in the synthesis of poly(2-vinylpyridine) (P2VP) by anionic polymerization. It can be industrially prepared by treatment of 2-methylpyridine with an aqueous solution of formaldehyde in a temperature range of 150°C to 200°C.Vinylation of silacyclobutanes with Ni-catalysis leading to geometrically pure (E)-silylolefins.

Specifications

CAS 100-69-6
Density 0.975 g/mL (at 25°C)
Boiling Point 79°C to 82°C (29 mmHg)
Molecular Formula C7H7N
Refractive Index n20/D 1.549 (literature)
Linear Formula C7H7N
Quantity 5 mL
MDL Number MFCD00006355
Synonym alpha-Vinylpyridine; 2-Ethenylpyridine; 2-Pyridylethylene
Molecular Weight (g/mol) 105.14
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.