Learn More
AVANTOR MASTERFLEX (VWR LLC) Two-Way Stopcock, HDPE, 14 mm hose barb, 7 psi max
Catalog No. NC1565264
Encompass
Chemical and heat resistant PVDF stopcocks allow you to stop flow quickly. Easily connect hose barb connectors to flexible tubing for quick and easy installation.
Catalog No. NC1565264 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9826120
Chemical and heat resistant PVDF stopcocks allow you to stop flow quickly. Easily connect hose barb connectors to flexible tubing for quick and easy installation.