Learn More
Thomas Scientific 2" White Freezer Box Only, No Drain Holes, 5 1/4" x 5 1/4" x 2"
SDP
Catalog No. NC0590356
Encompass_Preferred
Without Drain Holes, Premium Frost Resistant Box, Suitable for -80°C or Vapor Phase LN2
Catalog No. NC0590356 Supplier Thomas Scientific Supplier No. 1149H29
Without Drain Holes, Premium Frost Resistant Box, Suitable for -80°C or Vapor Phase LN2