Learn More
Gmp Labeling Inc Durable 2 x 1 ORNG Therm Trans Lbl
SDP
Catalog No. NC3483065
Encompass_Preferred
Roll Labels, Thermal Transfer, Chemical Resistant Polyester, 2" x 1", Orange, Permanent Ahdesive, 1000 labels/roll
Catalog No. NC3483065 Supplier Gmp Labeling Inc Supplier No. L221TTORG
Add to Cart
Add to Cart
Roll Labels, Thermal Transfer, Chemical Resistant Polyester, 2" x 1", Orange, Permanent Ahdesive, 1000 labels/roll