Learn More
Gmp Labeling Inc Durable 2 x 1 RED Therm Trans Lbl
SDP
Catalog No. NC3483068
Encompass_Preferred
Roll Labels, Thermal Transfer, Chemical Resistant Polyester, 2" x 1", Red, Permanent Ahdesive, 1000 labels/roll
Catalog No. NC3483068 Supplier Gmp Labeling Inc Supplier No. L221TTRED
Roll Labels, Thermal Transfer, Chemical Resistant Polyester, 2" x 1", Red, Permanent Ahdesive, 1000 labels/roll