Learn More
Gmp Labeling Inc Durable 2 x 1 YLLW Therm Trans Lbl
SDP
Catalog No. NC3483076
Encompass_Preferred
Roll Labels, Thermal Transfer, Chemical Resistant Polyester, 2" x 1", Yellow, Permanent Ahdesive, 1000 labels/roll
Catalog No. NC3483076 Supplier Gmp Labeling Inc Supplier No. L221TTYLW
Roll Labels, Thermal Transfer, Chemical Resistant Polyester, 2" x 1", Yellow, Permanent Ahdesive, 1000 labels/roll