Learn More
Gmp Labeling Inc Durable 2 x 2 GREEN Therm Trans Lbl
SDP
Catalog No. NC3483092
Encompass_Preferred
Roll Labels, Thermal Transfer, Chemical Resistant Polyester, 2" x 2", Green, Permanent Ahdesive, 1000 labels/roll
Catalog No. NC3483092 Supplier Gmp Labeling Inc Supplier No. L222TTGRN
Roll Labels, Thermal Transfer, Chemical Resistant Polyester, 2" x 2", Green, Permanent Ahdesive, 1000 labels/roll