Learn More
Chemglass Life Sciences Stir Bar, Micro, Magnetic, PTFE, Length x Dia. in mm: 2 x 2
SDP
Catalog No. CG200310
Encompass_Preferred
Stir Bar, Micro, Magnetic, PTFE, Length x Dia. in mm: 2 x 2. Micro PTFE stir bar for the very smallest containers. Manufactured with FDA-grade and USP Class VI PTFE.SEE CG-2003-50 FOR STIR BAR KIT
Catalog No. CG200310 Supplier Chemglass Life Sciences Supplier No. CG200310
Stir Bar, Micro, Magnetic, PTFE, Length x Dia. in mm: 2 x 2. Micro PTFE stir bar for the very smallest containers. Manufactured with FDA-grade and USP Class VI PTFE.SEE CG-2003-50 FOR STIR BAR KIT