Learn More
Gmp Labeling Inc Durable 2 x 2 WHITE Therm Trans Lbl
SDP
Catalog No. NC3483112
Encompass_Preferred
Roll Labels, Thermal Transfer, Chemical Resistant Polyester, 2" x 2", White, Permanent Ahdesive, 1000 labels/roll
Catalog No. NC3483112 Supplier Gmp Labeling Inc Supplier No. L222TTWHT
Roll Labels, Thermal Transfer, Chemical Resistant Polyester, 2" x 2", White, Permanent Ahdesive, 1000 labels/roll