Learn More
Gmp Labeling Inc Durable 2 x 3 ORNG Therm Trans Lbl
SDP
Catalog No. NC3483130
Encompass_Preferred
Roll Labels, Thermal Transfer, Chemical Resistant Polyester, 2" x 3", Orange, Permanent Ahdesive, 500 labels/roll
Catalog No. NC3483130 Supplier Gmp Labeling Inc Supplier No. L232TTORG
Roll Labels, Thermal Transfer, Chemical Resistant Polyester, 2" x 3", Orange, Permanent Ahdesive, 500 labels/roll