Learn More
Gmp Labeling Inc Durable 2 x 3 YLLW Therm Trans Lbl
SDP
Supplier:  Gmp Labeling Inc L232TTYLW
Encompass_Preferred
Roll Labels, Thermal Transfer, Chemical Resistant Polyester, 2" x 3", Yellow, Permanent Ahdesive, 500 labels/roll
Catalog No. NC3483160