Learn More
Thermo Scientific Viper Capillary MP35N, 0.100 mm x 750 mm
Supplier:  Thermo Scientific 6042.2390
Encompass
Viper Capillary MP35N, 0.100 mm x 750 mm, for use with all HPLC and UHPLC systems, 1500 bar maximum pressure
Catalog No. NC1045194