Learn More
Teknova 2XYT Broth. 1000mL, Sterile.
SDP
Supplier:  Teknova Y0167
Encompass
YP (Yeast Peptone) media is a growth medium used to culture yeast. YP is a rich source of proteins and nutrients essential for the development of yeast cells.
Catalog No. 50-843-437