Learn More
Teknova 2XYT Broth, 5kg.
SDP
Catalog No. 50843444
Encompass
1.6% Tryptone, 1.0% Yeast Extract, 0.5% Sodium Chloride
Catalog No. 50-843-444 Supplier Teknova Supplier No. Y0215
1.6% Tryptone, 1.0% Yeast Extract, 0.5% Sodium Chloride