Learn More
Medchemexpress LLC 2-[(Z)-(3,4-dibromophenyl)methylideneamino]guanidine | 849459-86-5 | 99.8% | 319.98 g/mol | C8H8Br2N4 | 100MG
SDP
Catalog No. 5000237152
Encompass_Preferred
AC-263093 free base is a research-grade small molecule described as a functionally selective neuropeptide FF receptor type 2 (NPFFR2) agonist. It is used in preclinical studies to investigate NPFF receptor signaling, pain modulation, and opioid tolerance. The compound has reported chemical formula C8H8Br2N4 and molecular weight ≈319.98 g/mol.
Catalog No. 50-002-37152 Supplier Medchemexpress LLC Supplier No. HY124680100MG
AC-263093 free base is a research-grade small molecule described as a functionally selective neuropeptide FF receptor type 2 (NPFFR2) agonist. It is used in preclinical studies to investigate NPFF receptor signaling, pain modulation, and opioid tolerance. The compound has reported chemical formula C8H8Br2N4 and molecular weight ≈319.98 g/mol.