Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 20:0-20:1-20:0 TG-d5 | 958764-14-2 | 99.9% | 978.65 | 1 MG
SDP

Catalog No. 5000362943
Encompass_Preferred

20:0-20:1-20:0 TG-d5 is deuterium labeled 20:0-20:1-20:0 TG. It is for research use only.

  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.

Catalog No. 50-003-62943 Supplier Medchemexpress LLC Supplier No. HY146917S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

20:0-20:1-20:0 TG-d5 is deuterium labeled 20:0-20:1-20:0 TG. It is for research use only.

  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.