Learn More
Thermco Products Inc Special application Non-Hg, 20/100C, .5C div, 76mm imm, 305mm
Catalog No. NC0227209
Encompass_Preferred
Precision spirit filled thermometer. The ACC711S thermometer was designed to be used in water baths with it's 76mm immersion. At the 76mm immersion there is an enlargement to assure of proper immersion Accurate to N.I.S.T. tolerances. Eligible for N.I.S.T. certification.
Catalog No. NC0227209 Supplier Thermco Products Inc Supplier No. ACC711S
Precision spirit filled thermometer. The ACC711S thermometer was designed to be used in water baths with it's 76mm immersion. At the 76mm immersion there is an enlargement to assure of proper immersion Accurate to N.I.S.T. tolerances. Eligible for N.I.S.T. certification.