Learn More
Electron Microscopy Sciences 20/40 Slide Glass Rack With Handle
SDP
Catalog No. 5030250
Encompass_Preferred
This glass rack holds 20 single slides or 40 slides back-to-back and is made from soda-lime glass.
Catalog No. 50-302-50 Supplier Electron Microscopy Sciences Supplier No. 7031236
This glass rack holds 20 single slides or 40 slides back-to-back and is made from soda-lime glass.