Learn More
Chemglass Life Sciences 20-400 G.P.I. Glass Thread, 19mm OD Tubing, 135mm OAL
SDP
Catalog No. CG19006
Encompass_Preferred
20-400 G.P.I. Glass Thread, 19mm OD Tubing, 135mm OAL. Thread for replacement on apparatus using G.P.I. (Glass Packaging Institute) thread.PLEASE NOTE: GPI THREADS ARE SOLD IN PACKAGES ONLY.
Catalog No. CG19006 Supplier Chemglass Life Sciences Supplier No. CG19006
20-400 G.P.I. Glass Thread, 19mm OD Tubing, 135mm OAL. Thread for replacement on apparatus using G.P.I. (Glass Packaging Institute) thread.PLEASE NOTE: GPI THREADS ARE SOLD IN PACKAGES ONLY.