Learn More
MSC Baldwin Filters Radial Seal Filter, 10-31/32x20-5/32 in, 1 ea
Catalog No. 5000504633
Encompass
These engine air intake filter elements protect heavy-duty equipment by preventing airborne dirt and other contaminants from entering the combustion chamber. Radial seal filters use a urethane compound to form molded ends and bond the media pack, creating a strong, yet flexible package. The heavy-duty radial seal gasket is designed for easier and more cost-effective servicing.
  • Protects engines and mobile equipment
  • High product quality
  • Supports rigorous applications
Catalog No. 50-005-04633 Supplier MSC Supplier No. 10948859
These engine air intake filter elements protect heavy-duty equipment by preventing airborne dirt and other contaminants from entering the combustion chamber. Radial seal filters use a urethane compound to form molded ends and bond the media pack, creating a strong, yet flexible package. The heavy-duty radial seal gasket is designed for easier and more cost-effective servicing.
  • Protects engines and mobile equipment
  • High product quality
  • Supports rigorous applications