Learn More
Waters Corp Waters Solvent Filter Assembly, 20 μL, 4/pk
Supplier:  Waters Corp 700001541
Encompass_Preferred
The 20μm Solvent Filter enables customers to replace the filter assembly on the solvent inlet tubes of the 2545 Binary Solvent Manager, 2545 Quaternary Solvent Manager, and 2555 Quaternary Solvent Manager.
  • Replaces filter assembly on solvent inlet tubes
  • Suitable for 2545 Binary Solvent Manager
  • Suitable for 2545 Quaternary Solvent Manager
  • Suitable for 2555 Quaternary Solvent Manager
  • Units per package: 4
  • Replacement schedule: As required
Catalog No. 50-203-9426