Learn More
WELCH MATERIALS INC DoprahTM Cap&Septa for Crimp Headspace Vial, 20mm Open Top Aluminum Crimp Cap (10mm hole) with 20mm Natural PTFE/White Silicone Septa 3mm Thick. 100pcs/pk, 10pk/cs.
Catalog No. NC2053898
Encompass_Preferred
Cap&Septa for Crimp Headspace Vial, 20mm Open Top Aluminum Crimp Cap (10mm hole) with 20mm Natural PTFE/White Silicone Septa 3mm Thick. 100pcs/pk, 10pk/cs.
Catalog No. NC2053898 Supplier WELCH MATERIALS INC Supplier No. DF119610
Cap&Septa for Crimp Headspace Vial, 20mm Open Top Aluminum Crimp Cap (10mm hole) with 20mm Natural PTFE/White Silicone Septa 3mm Thick. 100pcs/pk, 10pk/cs.