Learn More
MOLECULAR DEPOT LLC 20 Amino Acids Kit - 20/1 g
Catalog No. 5000013363
Encompass_Preferred
20 Amino Acids Kit (19 L amino acids plus Glycine) is a kit consisting of the 19 natural L-amino acids plus glycine Each amino acid is highly purified and is provided in one gram bottles This cost-effective kit is useful for amino acids screening reagents stabilization media formulation molecular labelling in vitro synthesis and other research applications For in vitro research use only
Catalog No. 50-000-13363 Supplier MOLECULAR DEPOT LLC Supplier No. B2010189
20 Amino Acids Kit (19 L amino acids plus Glycine) is a kit consisting of the 19 natural L-amino acids plus glycine Each amino acid is highly purified and is provided in one gram bottles This cost-effective kit is useful for amino acids screening reagents stabilization media formulation molecular labelling in vitro synthesis and other research applications For in vitro research use only