Learn More
National Safety Apparel™ Enespro™ 20 cal Arc Flash Faceshield Kit
SDP
Catalog No. 17355208 Shop All National Safety Apparel Products
Encompass_Preferred
Change view
Click to view available options
Arc Flash Rating:
20 cal/cm²
Catalog No. Arc Flash Rating
17-355-208 20 cal/cm²
1 options
This item is not returnable. View return policy
Catalog No. 17-355-208 Supplier National Safety Apparel™ Supplier No. KITHP20PV
This item is not returnable. View return policy
Arc flash faceshield kit includes a clear, low profile chin cup that increases field of vision and offers protection when looking down.
  • Premium Hover series bracket for weight compensation over hard hat
  • Highly transparent arc flash faceshield for greater visibility
  • Proven color recognition
  • Low profile chin cup
  • Anti-fog and scratch resistant coating
  • Adjustable settings allow the faceshield to be secured in various positions
  • Lightweight design
Specifications
Product Type Arc Flash Head Protection Kit
Size Universal
Material Plastic, UltraSoft
Color Clear
Disposable Reusable
Arc Flash Rating 20 cal/cm²
Certifications/Compliance NFPA 70E, ASTM F1506, ASTM F2178, CSA Z462, ANSI/ISEA Z87.1, OSHA 1910.269
Quantity 1 Ea.
Includes 20 cal lift front faceshield, balaclava, and bag
Style Faceshield Kit