Learn More
National Safety Apparel™ 20 Cal Ultrasoft Arc Flash Coat
SDP
Catalog No. 17001269 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-269 Small
17-001-270 Medium
17-001-271 Large
17-001-272 2X-Large
17-001-273 3X-Large
17-001-274 X-Large
17-001-275 4X-Large
17-001-276 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-001-269 Supplier National Safety Apparel™ Supplier No. C04UW03SM32
Add to Cart
Add to Cart
For enhanced protection Westex™ Ultrasoft is the ideal choice for superior comfort, improved safety, and above average durability
  • Arc Rating: 20 cal/cm²
  • CAT 2
  • Fabric/Material: 13 oz. Westex™ Ultrasoft
  • Breathable FR fabric
  • FR fabric will not melt, drip or ignite
  • FR knit wrist cuffs keep sleeves in place
  • Generous fit for ease of movement
  • Inherently flame resistant protection will not wash/wear out
  • Hook and loop front closure
  • Made in the USA
Specifications
Closure Type Hook and Loop
Color Navy
Disposable Reusable
Gender Unisex
Material Cotton, Nylon
Neck Style Collared
Size Small
Resistance Flame
Seam Type Stitched
Arc Flash Rating 20 cal/cm²
Certifications/Compliance CAT 2, ASTM F1506, OSHA 1910.269, NFPA 70 E, CSA Z462