Learn More
JDM LAB SOLUTIONS LLC 20° Digital Nutating Mixer NS-11UA
Catalog No. NC2385881
Encompass
Tilt angles are pre-set for 3D motionAll models come with a dimpled non-skid mat.Compact and economic choice for 3D mixing (Models NS-01A and NS-10A)Variable speed from 2-60 rpm (Models NS-11UA and NS-12UA models)Continuous or time set operation with an automatic switch-off feature (Models NS-11UA and NS-12UA models).Audible and visual alarm in time mode feature (Models NS-11UA and NS-12UA models).
Catalog No. NC2385881 Supplier JDM LAB SOLUTIONS LLC Supplier No. NS11UA
Tilt angles are pre-set for 3D motionAll models come with a dimpled non-skid mat.Compact and economic choice for 3D mixing (Models NS-01A and NS-10A)Variable speed from 2-60 rpm (Models NS-11UA and NS-12UA models)Continuous or time set operation with an automatic switch-off feature (Models NS-11UA and NS-12UA models).Audible and visual alarm in time mode feature (Models NS-11UA and NS-12UA models).