Learn More
Description
- Used in extremely stable environments
- 1 to 500 mg weights are one-piece construction with no adjusting cavity with one end turned up for easy handling
- Weights 1 g and above are of two-piece construction
- Weight sets 1 mg-500 g are provided in a high-quality polycarbonate set case
Specifications
Specifications
| Certifications/Compliance | Traceable Certificate |
| Classification | UltraClass |
| Material | Troemner Alloy 8, Stainless Steel |
| Case Material | Polycarbonate |
| Density | Weights 1 g and above are 8.03 g/cm3 at 20°C. Weights below 1 g are 7.95 g/cm3 at 20°C. |
| Mass | 20 g to 100 mg |
| Length (Metric) | 20.32 cm |
| Height (Metric) Case | 8.89 cm |
| For Use With (Application) | Reference standard calibration |
| For Use With (Equipment) | Calibration of high-precision analytical balances with readability as low as 0.01 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.